Benzyl isocyanide ≥96.0%
Supplier: TCI America
B2185-1G
B2185-5G
TCB2185-1GEA
90.22
USD
TCB2185-1G
TCB2185-5G
Benzyl isocyanide ≥96.0%
Benzyl isocyanide
CAS Number: 10340-91-7
MDL Number: MFCD00000004
Molecular Formula: C8H7N
Molecular Weight: 117.15
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 79
Specific Gravity (20/20): 0.99
Storage Temperature: <0°C
Formula:
C₈H₇N MW: 117.15 g/mol Boiling Pt: 106 °C Density: 0.99 (20 °C) Flash Pt: 79 °C |
MDL Number:
MFCD00000004 CAS Number: 10340-91-7 UN: 2810 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...