3-Picolyl bromide hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2170-25G B2170-5G
TCB2170-25GEA 649 USD
TCB2170-25G TCB2170-5G
3-Picolyl bromide hydrobromide ≥98.0% (by titrimetric analysis)
3-Picolyl bromide hydrobromide

CAS Number: 4916-55-6
MDL Number: MFCD01321314
Molecular Formula: C6H6BrN
Molecular Weight: 252.94
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 150


Other Product Names / Synonyms: 3-Picolyl Bromide Hydrobromide

Formula: C₆H₇Br₂N
Melting Pt: 150 °C
MDL Number: MFCD01321314
CAS Number: 4916-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38bd1e1efc454250b0c0e3d23e2465775cdeb4f5&vid=38bd1e1efc454250b0c0e3d23e2465775cdeb4f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR