Bis(dimethylamino)dimethylsilane ≥97.0%

Supplier: TCI America

B2150-250ML B2150-25ML
TCB2150-250MLEA 284 USD
TCB2150-250ML TCB2150-25ML
Bis(dimethylamino)dimethylsilane ≥97.0%
Bis(dimethylamino)dimethylsilane

CAS Number: 3768-58-9
MDL Number: MFCD00008296
Molecular Formula: C6H18N2Si
Molecular Weight: 146.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Flash Point (°C): -7
Specific Gravity (20/20): 0.81

Formula: C₆H₁₈N₂Si
Boiling Pt: 128 °C
Density: 0.81 (20 °C)
Flash Pt: -7 °C
MDL Number: MFCD00008296
CAS Number: 3768-58-9
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2579e2a13d55841f64041503a58817ed00b1c54b&vid=2579e2a13d55841f64041503a58817ed00b1c54b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR