2-Bromophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-Bromophenylhydrazine hydrochloride, 1-(2-Bromophenyl)hydrazine hydrochloride
B2147-25G
B2147-5G
TCB2147-25GEA
274
USD
TCB2147-25G
TCB2147-5G
2-Bromophenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
2-Bromophenylhydrazinium chloride
CAS Number: 50709-33-6
MDL Number: MFCD00012926
Molecular Formula: C6H7BrN2
Molecular Weight: 223.50
Purity/Analysis Method:
Formula:
C₆H₈BrClN₂ MW: 223.5 g/mol Melting Pt: 185 °C |
MDL Number:
MFCD00012926 CAS Number: 50709-33-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...