ω-Bromo-4-methylacetophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

B2107-25G B2107-5G
TCB2107-25GEA 77.33 USD
TCB2107-25G TCB2107-5G
ω-Bromo-4-methylacetophenone ≥98.0% (by GC)
ω-Bromo-4-methylacetophenone

CAS Number: 619-41-0
MDL Number: MFCD00000203
Molecular Formula: C9H9BrO
Molecular Weight: 213.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 53
Flash Point (°C): 110


Other Product Names / Synonyms: 4-(Bromoacetyl)toluene, 4-Methylphenacyl Bromide

Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 159 °C
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD00000203
CAS Number: 619-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=738c243b875b6e5f93cc7e3c4a7aef84c6a3788e&vid=738c243b875b6e5f93cc7e3c4a7aef84c6a3788e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR