6-Bromo-1-hexene ≥95.0% (by GC)
Supplier: TCI America
B2106-25G
B2106-5G
TCB2106-25GEA
182.99
USD
TCB2106-25G
TCB2106-5G
6-Bromo-1-hexene ≥95.0% (by GC)
6-Bromo-1-hexene
CAS Number: 2695-47-8
MDL Number: MFCD00000269
Molecular Formula: C6H11Br
Molecular Weight: 163.06
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 54
Specific Gravity (20/20): 1.22
Storage Temperature: <0°C
Other Product Names / Synonyms: 5-Hexenyl Bromide
Formula:
C₆H₁₁Br MW: 163.06 g/mol Boiling Pt: 147 °C Density: 1.22 (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00000269 CAS Number: 2695-47-8 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...