5-Bromovaleryl chloride ≥98.0%
Supplier: TCI America
Synonyms:
5-Bromopentanoyl chloride, 5-Bromovalerylchloride
B2059-250G
B2059-25G
B2059-5G
TCB2059-250GEA
486
USD
TCB2059-250G
TCB2059-25G
TCB2059-5G
5-Bromovaleryl chloride ≥98.0%
5-Bromovaleryl chloride
CAS Number: 4509-90-4
MDL Number: MFCD00013660
Molecular Formula: C5H8BrClO
Molecular Weight: 199.47
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 107
Specific Gravity (20/20): 1.51
Other Product Names / Synonyms: 5-Bromovaleroyl Chloride
Formula:
BrH₂C(CH₂)₃COOCl MW: 199.47 Boiling Pt: 118 °C Density: 1.51 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00013660 CAS Number: 4509-90-4 UN: 3265 ADR: 8 II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...