3,5-Bis(trifluoromethyl)benzoyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,5-Bis(trifluoromethyl)benzene-1-carbonyl chloride, 3,5-bis(trifluoromethyl)benzoylchloride

B2000-25G B2000-5G
TCB2000-25GEA 74.25 USD
TCB2000-25G TCB2000-5G
3,5-Bis(trifluoromethyl)benzoyl chloride ≥98.0% (by titrimetric analysis)
3,5-Bis(trifluoromethyl)benzoyl chloride

CAS Number: 785-56-8
MDL Number: MFCD00000387
Molecular Formula: C9H3ClF6O
Molecular Weight: 276.56
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 72
Specific Gravity (20/20): 1.54

Formula: C₉H₃ClF₆O
MW: 276.57 g/mol
Boiling Pt: 81 °C
Density: 1.54 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00000387
CAS Number: 785-56-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc4915fab89f6c436790ef066fd12dd94b7046ae&vid=cc4915fab89f6c436790ef066fd12dd94b7046ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR