5-Bromo-2-furaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

B1954-25G B1954-5G
TCB1954-25GEA 156 USD
TCB1954-25G TCB1954-5G
5-Bromo-2-furaldehyde ≥98.0% (by GC)
5-Bromo-2-furaldehyde

CAS Number: 1899-24-7
MDL Number: MFCD00159501
Molecular Formula: C5H3BrO2
Molecular Weight: 174.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 84
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Bromofurfural

Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Boiling Pt: 112 °C
Melting Pt: 84 °C
MDL Number: MFCD00159501
CAS Number: 1899-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbaf42871bfdbda09c76b08fae3157bc0c1ef9d8&vid=dbaf42871bfdbda09c76b08fae3157bc0c1ef9d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR