4-Bromo-3-fluorotoluene ≥94.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-2-fluoro-4-methylbenzene

B1935-25G B1935-5G
TCB1935-25GEA 122.4 USD
TCB1935-25G TCB1935-5G
4-Bromo-3-fluorotoluene ≥94.0% (by GC)
4-Bromo-3-fluorotoluene

CAS Number: 452-74-4
MDL Number: MFCD00040940
Molecular Formula: C7H6BrF
Molecular Weight: 189.03
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 191
Flash Point (°C): 35
Specific Gravity (20/20): 1.51

Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 191 °C
Density: 1.51 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00040940
CAS Number: 452-74-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aeee5d50fbcddbf5cdf6b6b2d0055676c837e0d1&vid=aeee5d50fbcddbf5cdf6b6b2d0055676c837e0d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR