α,4-Dibromo-2-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromo-1-(bromomethyl)-2-fluorobenzene, 4-Bromo-2-fluorobenzyl bromide

B1934-25G B1934-5G
TCB1934-25GEA 97 USD
TCB1934-25G TCB1934-5G
α,4-Dibromo-2-fluorotoluene ≥98.0% (by GC)
α,4-Dibromo-2-fluorotoluene

CAS Number: 76283-09-5
MDL Number: MFCD00055467
Molecular Formula: C7H5Br2F
Molecular Weight: 267.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 37

Formula: C₇H₅Br₂F
MW: 267.92 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00055467
CAS Number: 76283-09-5
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5382a8aa24b7ece1ede510687807635ceecce64&vid=131da1f79309991a6200897ddc9b3a60e530b518&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR