3-Bromopropionaldehyde dimethylacetal ≥98.0% (by GC)
Supplier: TCI America
B1901-25G
B1901-5G
TCB1901-25GEA
658
USD
TCB1901-25G
TCB1901-5G
3-Bromopropionaldehyde dimethylacetal ≥98.0% (by GC)
3-Bromopropionaldehyde dimethylacetal
CAS Number: 36255-44-4
MDL Number: MFCD00013245
Molecular Formula: C5H11BrO2
Molecular Weight: 183.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 46
Specific Gravity (20/20): 1.36
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 1-Bromo-3,3-dimethoxypropane
Formula:
BrH₂CCH₂CH(OCH₃)₂ Boiling Pt: 60 °C Density: 1.36 (20 °C) Flash Pt: 46 °C |
MDL Number:
MFCD00013245 CAS Number: 36255-44-4 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...