3,5-Bis(trifluoromethyl)benzyl bromide ≥96.0%

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene

B1867-25G B1867-5G
TCB1867-25GEA 147 USD
TCB1867-25G TCB1867-5G
3,5-Bis(trifluoromethyl)benzyl bromide ≥96.0%
3,5-Bis(trifluoromethyl)benzyl bromide

CAS Number: 32247-96-4
MDL Number: MFCD00009905
Molecular Formula: C9H5BrF6
Molecular Weight: 307.03
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 26
Freezing point (°C): 18
Specific Gravity (20/20): 1.67


Other Product Names / Synonyms: 1-Bromomethyl-3,5-bis(trifluoromethyl)benzene

Formula: C₉H₅BrF₆
Boiling Pt: 140 °C
Density: 1.67 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00009905
CAS Number: 32247-96-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ad100a64d1a2269c7aec7927e8d2a3963cc4f5c&vid=8ad100a64d1a2269c7aec7927e8d2a3963cc4f5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR