1-Bromo-3-chloro-4-fluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Bromo-2-chloro-1-fluorobenzene, 4-Bromo-2-chlorofluorobenzene

B1860-25G B1860-5G
TCB1860-25GEA 60.7 USD
TCB1860-25G TCB1860-5G
1-Bromo-3-chloro-4-fluorobenzene ≥98.0% (by GC)
1-Bromo-3-chloro-4-fluorobenzene

CAS Number: 60811-21-4
MDL Number: MFCD00051794
Molecular Formula: C6H3BrClF
Molecular Weight: 209.44
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 88
Specific Gravity (20/20): 1.74

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 95 °C
Density: 1.74 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00051794
CAS Number: 60811-21-4
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8167bd867b54775c84b131652732dab163f8dd7a&vid=b12681de55dbacb300f7c180a3095246a1575e2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR