1-Bromo-3,4,5-trifluorobenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5-Bromo-1,2,3-trifluorobenzene
B1765-25G
B1765-5G
TCB1765-25GEA
48
USD
TCB1765-25G
TCB1765-5G
1-Bromo-3,4,5-trifluorobenzene ≥98.0% (by GC)
1-Bromo-3,4,5-trifluorobenzene
CAS Number: 138526-69-9
MDL Number: MFCD00070738
Molecular Formula: C6H2BrF3
Molecular Weight: 210.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 43
Flash Point (°C): 45
Specific Gravity (20/20): 1.78
Formula:
C₆H₂BrF₃ MW: 210.98 g/mol Boiling Pt: 43 °C Density: 1.78 (20 °C) Flash Pt: 45 °C |
MDL Number:
MFCD00070738 CAS Number: 138526-69-9 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...