2,4-Difluorobenzyl bromide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene

B1710-25G B1710-5G
TCB1710-25GEA 223 USD
TCB1710-25G TCB1710-5G
2,4-Difluorobenzyl bromide ≥98.0% (by GC)
2,4-Difluorobenzyl bromide

CAS Number: 23915-07-3
MDL Number: MFCD00011649
Molecular Formula: C7H5BrF2
Molecular Weight: 207.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 40
Freezing point (°C): 18
Specific Gravity (20/20): 1.63


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-2,4-difluorotoluene

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Density: 1.63 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00011649
CAS Number: 23915-07-3
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16062775582e5f5aadc1e96eba58804950ad673e&vid=5b1cbe97fab1a37930fc73411e6312b0fe642d1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR