9-Bromo-9-phenylfluorene ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 9-Bromo-9-phenyl-9H-fluorene

B1702-25G B1702-5G
TCB1702-25GEA 341 USD
TCB1702-25G TCB1702-5G
9-Bromo-9-phenylfluorene ≥96.0% (by GC)
9-Bromo-9-phenylfluorene

CAS Number: 55135-66-5
MDL Number: MFCD00075522
Molecular Formula: C19H13Br
Molecular Weight: 321.22
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 100

Formula: C₁₉H₁₃Br
MW: 321.23 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00075522
CAS Number: 55135-66-5
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62389385f6a401fc7bec8178413d1396c58f3704&vid=62389385f6a401fc7bec8178413d1396c58f3704&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR