Bis(acetonitrile)palladium(II) dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bis(acetonitrile)-palladium(II) chloride, Palladium(II) chloride diacetonitrile complex, Dichlorobis(acetonitrile)palladium(II)

B1676-1G B1676-5G
TCB1676-1GEA 291.9 USD
TCB1676-1G TCB1676-5G
Bis(acetonitrile)palladium(II) dichloride ≥98.0% (by titrimetric analysis)
Bis(acetonitrile)palladium(II) dichloride

CAS Number: 14592-56-4
MDL Number: MFCD00013122
Molecular Formula: C4H6Cl2N2Pd
Molecular Weight: 259.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction
Other Product Names / Synonyms: Dichlorobis(acetonitrile)palladium(II)

Formula: C₄H₆Cl₂N₂Pd
MW: 259.43 g/mol
MDL Number: MFCD00013122
CAS Number: 14592-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=308bc97b42b77fb2d2dbaa308d048c1336df803d&vid=308bc97b42b77fb2d2dbaa308d048c1336df803d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR