4'-tert-Butylpropiophenone ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 1-[4-(1,1-Dimethylethyl)phenyl]-1-propanone

B1609-1ML B1609-5ML
TCB1609-1MLEA 91 USD
TCB1609-1ML TCB1609-5ML
4'-tert-Butylpropiophenone ≥95.0% (by GC)
4'-tert-Butylpropiophenone

CAS Number: 71209-71-7
MDL Number: MFCD00100434
Molecular Formula: C13H18O
Molecular Weight: 190.29
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 4-tert-Butylpropionylbenzene

Formula: C₁₃H₁₈O
MW: 190.28 g/mol
Boiling Pt: 162 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00100434
CAS Number: 71209-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=487a52e59ae2985b7bb31819f339b7cd936ce801&vid=487a52e59ae2985b7bb31819f339b7cd936ce801&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR