Butyraldehyde-2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B1594-10G
B1594-1G
TCB1594-10GEA
260
USD
TCB1594-10G
TCB1594-1G
Butyraldehyde-2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Butyraldehyde-2,4-dinitrophenylhydrazone
CAS Number: 1527-98-6
MDL Number: MFCD00191327
Molecular Formula: C10H12N4O4
Molecular Weight: 252.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 122
Formula:
C₁₀H₁₂N₄O₄ MW: 252.23 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00191327 CAS Number: 1527-98-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...