4-tert-Butylbenzylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
p-tert-Butylbenzylamine
B1587-25ML
B1587-5ML
TCB1587-25MLEA
163
USD
TCB1587-25ML
TCB1587-5ML
4-tert-Butylbenzylamine ≥98.0% (by GC, titration analysis)
4-tert-Butylbenzylamine
CAS Number: 39895-55-1
MDL Number: MFCD00040754
Molecular Formula: C11H17N
Molecular Weight: 163.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 107
Specific Gravity (20/20): 0.93
Formula:
C₁₁H₁₇N MW: 163.26 g/mol Boiling Pt: 124 °C Density: 0.93 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00040754 CAS Number: 39895-55-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...