4-tert-Butylbenzonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 4210-32-6
MDL Number: MFCD00075840
Molecular Formula: C11H13N
Molecular Weight: 159.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 110
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 4-tert-Butylcyanobenzene
Formula:
C₁₁H₁₃N Boiling Pt: 112 °C Density: 0.95 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00075840 CAS Number: 4210-32-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...