α,3-Dibromotoluene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene
B1570-25G
B1570-5G
TCB1570-25GEA
41
USD
TCB1570-25G
TCB1570-5G
α,3-Dibromotoluene ≥98.0% (by GC)
α,3-Dibromotoluene
CAS Number: 823-78-9
MDL Number: MFCD00000176
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 42
Flash Point (°C): 110
Storage Temperature: 0-10°C
Other Product Names / Synonyms: alpha,3-Dibromotoluene
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Boiling Pt: 135 °C Melting Pt: 42 °C Flash Pt: 110 °C |
MDL Number:
MFCD00000176 CAS Number: 823-78-9 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...