2-Bromo-4-fluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1555-25G B1555-5G
TCB1555-25GEA 82 USD
TCB1555-25G TCB1555-5G
2-Bromo-4-fluorophenol ≥98.0% (by GC, titration analysis)
2-Bromo-4-fluorophenol

CAS Number: 496-69-5
MDL Number: MFCD00010614
Molecular Formula: C6H4BrFO
Molecular Weight: 191.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 89
Melting point (°C): 43

Formula: C₆H₄BrFO
MW: 191 g/mol
Boiling Pt: 89 °C
Melting Pt: 43 °C
MDL Number: MFCD00010614
CAS Number: 496-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f652e9c182cb58b550e6d6f5af023ddfbcfbee0d&vid=4c83f476017ee58fbc6c73a0c71b214e5edd02ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR