1,3-Bis(3-aminopropyl)tetramethyldisiloxane ≥83.0% (by GC)
Supplier: TCI America
[Monomer for silicon modified polyamides]
CAS Number: 2469-55-8
MDL Number: MFCD00039786
Molecular Formula: C10H28N2OSi2
Molecular Weight: 248.52
Purity/Analysis Method: >83.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 91
Specific Gravity (20/20): 0.90
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Tetramethyl-1,3-bis(3-aminopropyl)disiloxane
Formula:
C₁₀H₂₈N₂OSi₂ Boiling Pt: 142 °C Density: 0.90 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00039786 CAS Number: 2469-55-8 |
|
Specification Test Results
Purity/Analysis Method | >83.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...