2-(Bromomethyl)naphthalene ≥97.0% (by GC)

Supplier: TCI America

B1524-25G B1524-5G
TCB1524-25GEA 373.5 USD
TCB1524-25G TCB1524-5G
2-(Bromomethyl)naphthalene ≥97.0% (by GC)
2-(Bromomethyl)naphthalene

CAS Number: 939-26-4
Molecular Formula: C11H9Br
Molecular Weight: 221.10
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 214
Melting point (°C): 55
Flash Point (°C): 110

Formula: C₁₁H₉Br
MW: 221.1 g/mol
Boiling Pt: 214 °C
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00004123
CAS Number: 939-26-4
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcda7d20b95f85a6b505a0cdcd0cbce9d0ffd24a&vid=fcda7d20b95f85a6b505a0cdcd0cbce9d0ffd24a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR