4-Bromo-1,2-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-3,4-difluorobenzene

B1523-250G B1523-25G
TCB1523-250GEA 182 USD
TCB1523-250G TCB1523-25G
4-Bromo-1,2-difluorobenzene ≥98.0% (by GC)
4-Bromo-1,2-difluorobenzene

CAS Number: 348-61-8
MDL Number: MFCD00000304
Molecular Formula: C6H3BrF2
Molecular Weight: 192.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 33
Specific Gravity (20/20): 1.71


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3,4-Difluorobromobenzene

Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 151 °C
Density: 1.71 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00000304
CAS Number: 348-61-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=921509fa0d7ed1d60bde8dcffcc1cc26fe9052a8&vid=b40cfb305f91929a7120b701712a5d33187aa12d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR