tert-Butyl trichloroacetimidate ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
tert-Butyl 2,2,2-trichloroacetimidate, t-Butyl trichioroacetimidate, t-butyl-2,2,2-trichloroacetimidate
CAS Number: 98946-18-0
MDL Number: MFCD00077410
Molecular Formula: C6H10Cl3NO
Molecular Weight: 218.50
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 69
Flash Point (°C): 55
Freezing point (°C): 20
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 2,2,2-Trichloroacetimidic Acid tert-Butyl Ester
Formula:
C₆H₁₀Cl₃NO MW: 218.502 g/mol Boiling Pt: 69 °C Density: 1.22 (20 °C) Flash Pt: 55 °C |
MDL Number:
MFCD00077410 CAS Number: 98946-18-0 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...