tert-Butyl methyl sulfide ≥98.0% (by GC)

Supplier: TCI America

B1475-25ML B1475-5ML
TCB1475-25MLEA 100.7 USD
TCB1475-25ML TCB1475-5ML
tert-Butyl methyl sulfide ≥98.0% (by GC)
tert-Butyl methyl sulfide

CAS Number: 6163-64-0
MDL Number: MFCD00008840
Molecular Formula: C5H12S
Molecular Weight: 104.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): -3
Specific Gravity (20/20): 0.83

Formula: C₅H₁₂S
MW: 104.21 g/mol
Boiling Pt: 102 °C
Density: 0.83 (20 °C)
Flash Pt: –3 °C
MDL Number: MFCD00008840
CAS Number: 6163-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a406c14174375a9e93cdebe3b4bfc20f8d80cb6b&vid=a406c14174375a9e93cdebe3b4bfc20f8d80cb6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR