5-Bromo-1-pentene ≥95.0% (by GC)

Supplier: TCI America

B1474-25G B1474-5G
TCB1474-25GEA 103.6 USD
TCB1474-25G TCB1474-5G
5-Bromo-1-pentene ≥95.0% (by GC)
5-Bromo-1-pentene

CAS Number: 1119-51-3
MDL Number: MFCD00000264
Molecular Formula: C5H9Br
Molecular Weight: 149.03
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 30
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Pentenyl Bromide

Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 126 °C
Density: 1.26 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00000264
CAS Number: 1119-51-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=387f22e2651c1d0e0973e2082877168805a84cf7&vid=d480d4dced4390896b9cf0db2a1c46555b39a82f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR