Bis(trimethylsilyl)carbodiimide ≥98.0% (by GC)

Supplier: TCI America

B1368-25G B1368-5G
TCB1368-25GEA 312 USD
TCB1368-25G TCB1368-5G
Bis(trimethylsilyl)carbodiimide ≥98.0% (by GC)
Bis(trimethylsilyl)carbodiimide

CAS Number: 1000-70-0
MDL Number: MFCD00051538
Molecular Formula: C7H18N2Si2
Molecular Weight: 186.41
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 41
Specific Gravity (20/20): 0.83

Formula: C₇H₁₈N₂Si₂
MW: 186.4 g/mol
Boiling Pt: 164 °C
Density: 0.83 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00051538
CAS Number: 1000-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09b52f8d644398e3d846e6778255dda102861617&vid=09b52f8d644398e3d846e6778255dda102861617&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR