2-Bromo-5-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

B1303-25G B1303-5G
TCB1303-25GEA 34 USD
TCB1303-25G TCB1303-5G
2-Bromo-5-fluorotoluene ≥98.0% (by GC)
2-Bromo-5-fluorotoluene

CAS Number: 452-63-1
MDL Number: MFCD00017921
Molecular Formula: C7H6BrF
Molecular Weight: 189.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 45
Specific Gravity (20/20): 1.53

Formula: C₇H₆BrF
Boiling Pt: 177 °C
Density: 1.53 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00017921
CAS Number: 452-63-1
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3735fd91d6a41fe6f3bf2d3dc0545f1bf9e803b&vid=007b914c685894ebab66184cbf1fa70802791044&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR