6-Bromocaproic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B1290-250G
B1290-25G
TCB1290-250GEA
373.64
USD
TCB1290-250G
TCB1290-25G
6-Bromocaproic acid ≥98.0% (by GC, titration analysis)
6-Bromocaproic acid
CAS Number: 4224-70-8
MDL Number: MFCD00004422
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 35
Flash Point (°C): 95
Storage Temperature: 0-10°C
Formula:
BrH₂C(CH₂)₄COOH Boiling Pt: 170 °C Melting Pt: 35 °C Flash Pt: 95 °C |
MDL Number:
MFCD00004422 CAS Number: 4224-70-8 UN: 3261 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...