(4-Biphenyl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Biphenylacetic acid

B1278-250G B1278-25G
TCB1278-250GEA 233 USD
TCB1278-250G TCB1278-25G
(4-Biphenyl)acetic acid ≥98.0%
(4-Biphenyl)acetic acid

CAS Number: 5728-52-9
MDL Number: MFCD00004351
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 163


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: Felbinac

Formula: (C₆H₅C₆H₅)CH₂COOH
Melting Pt: 163 °C
MDL Number: MFCD00004351
CAS Number: 5728-52-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70a1a7a6e3648963ae602ef107ae31299da714c0&vid=70a1a7a6e3648963ae602ef107ae31299da714c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR