2-(Bromoacetyl)naphthalene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone
B1259-25G
B1259-5G
TCB1259-25GEA
86
USD
TCB1259-25G
TCB1259-5G
2-(Bromoacetyl)naphthalene ≥98.0% (by GC)
2-(Bromoacetyl)naphthalene
CAS Number: 613-54-7
MDL Number: MFCD00004109
Molecular Formula: C12H9BrO
Molecular Weight: 249.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84
Other Product Names / Synonyms: 2-Bromo-2'-acetonaphthone, Bromomethyl 2-Naphthyl Ketone
Formula:
C₁₂H₉BrO MW: 249.11 g/mol Melting Pt: 84 °C |
MDL Number:
MFCD00004109 CAS Number: 613-54-7 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...