3-Bromo-1,1,1-trifluoroacetone ≥95.0% (by GC)

Supplier: TCI America

B1240-25G B1240-5G
TCB1240-25GEA 106.08 USD
TCB1240-25G TCB1240-5G
3-Bromo-1,1,1-trifluoroacetone ≥95.0% (by GC)
3-Bromo-1,1,1-trifluoroacetone

CAS Number: 431-35-6
MDL Number: MFCD00039237
Molecular Formula: C3H2BrF3O
Molecular Weight: 190.95
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 5
Specific Gravity (20/20): 1.82


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Bromo-1,1,1-trifluoroacetone, 1-Bromo-3,3,3-trifluoro-2-propanone, 3-Bromo-1,1,1-trifluoro-2-propanone

Formula: BrH₂CC(O)CF₃
MW: 190.95 g/mol
Boiling Pt: 88 °C
Density: 1.82 (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00039237
CAS Number: 431-35-6
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6593ed9b806da411ecfe07c8a46e08c3e66c8db5&vid=b2f3b892dd0df8c0f152480ad3c6cdffe20a12f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR