2-Bromobenzoyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B1238-250G B1238-25G
TCB1238-250GEA 579 USD
TCB1238-250G TCB1238-25G
2-Bromobenzoyl chloride ≥98.0% (by GC, titration analysis)
2-Bromobenzoyl chloride

CAS Number: 7154-66-7
MDL Number: MFCD00000655
Molecular Formula: C7H4BrClO
Molecular Weight: 219.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 110
Freezing point (°C): 10
Specific Gravity (20/20): 1.68

Formula: C₇H₄BrClO
Boiling Pt: 245 °C
Density: 1.68 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000655
CAS Number: 7154-66-7
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13d778447b7a346537a64e20c9e6369679ac4042&vid=13d778447b7a346537a64e20c9e6369679ac4042&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR