tert-Butylacetyl chloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,3-Dimethylbutyryl chloride

B1196-250ML B1196-25ML
TCB1196-250MLEA 327.4 USD
TCB1196-250ML TCB1196-25ML
tert-Butylacetyl chloride ≥99.0% (by titrimetric analysis)
tert-Butylacetyl chloride

CAS Number: 7065-46-5
MDL Number: MFCD00000737
Molecular Formula: C6H11ClO
Molecular Weight: 134.60
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 21
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: tert-Butylacetyl Chloride

Formula: C₆H₁₁ClO
MW: 134.61
Boiling Pt: 132 °C
Density: 0.97 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00000737
CAS Number: 7065-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee4484562913d07b3f462827d3286f6f12e23ca3&vid=ee4484562913d07b3f462827d3286f6f12e23ca3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR