1-Bromo-2,4,6-trifluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-1,3,5-trifluorobenzene

B1168-25G B1168-5G
TCB1168-25GEA 266 USD
TCB1168-25G TCB1168-5G
1-Bromo-2,4,6-trifluorobenzene ≥98.0% (by GC)
1-Bromo-2,4,6-trifluorobenzene

CAS Number: 2367-76-2
MDL Number: MFCD00000334
Molecular Formula: C6H2BrF3
Molecular Weight: 210.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Melting point (°C): 4
Flash Point (°C): 58
Specific Gravity (20/20): 1.80

Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 140 °C
Melting Pt: 4 °C
Density: 1.80 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00000334
CAS Number: 2367-76-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5680e6379dd1d5346ccebc805a64999050ed2a58&vid=1187cb8fadee49ba8c8103cd6da4e6be68a9ea06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR