1-Bromo-2,4,5-trifluorobenzene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2,4,5-Trifluorobromobenzene
B1167-25G
B1167-5G
TCB1167-25GEA
81.6
USD
TCB1167-25G
TCB1167-5G
1-Bromo-2,4,5-trifluorobenzene ≥98.0% (by GC)
1-Bromo-2,4,5-trifluorobenzene
CAS Number: 327-52-6
MDL Number: MFCD00000306
Molecular Formula: C6H2BrF3
Molecular Weight: 210.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 55
Specific Gravity (20/20): 1.81
Formula:
C₆H₂BrF₃ MW: 210.98 g/mol Boiling Pt: 144 °C Density: 1.81 (20 °C) Flash Pt: 55 °C |
MDL Number:
MFCD00000306 CAS Number: 327-52-6 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...