Bromopyruvic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid
B1153-25G
B1153-5G
TCB1153-25GEA
321
USD
TCB1153-25G
TCB1153-5G
Bromopyruvic acid ≥98.0% (by titrimetric analysis)
Bromopyruvic acid
CAS Number: 1113-59-3
MDL Number: MFCD00002587
Molecular Formula: C3H3BrO3
Molecular Weight: 166.96
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 77
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Bromo-2-oxopropionic Acid
Formula:
BrH₂CC(O)COOH MW: 166.96 g/mol Melting Pt: 77 °C |
MDL Number:
MFCD00002587 CAS Number: 1113-59-3 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...