1,2-Benzenedisulfonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Benzene-1,2-di(sulphonyl chloride)

B1128-1G B1128-25G B1128-5G
TCB1128-1GEA 193 USD
TCB1128-1G TCB1128-25G TCB1128-5G
1,2-Benzenedisulfonyl chloride ≥98.0% (by titrimetric analysis)
1,2-Benzenedisulphonyl chloride

CAS Number: 6461-76-3
MDL Number: MFCD00059836
Molecular Formula: C6H4Cl2O4S2
Molecular Weight: 275.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 144


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: 1,2-Benzenedisulfonyl Chloride

Formula: C₆H₄Cl₂O₄S₂
MW: 275.13 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00059836
CAS Number: 6461-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a73a2cece766d2aa45d84d30efe63066a5e00716&vid=a73a2cece766d2aa45d84d30efe63066a5e00716&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR