Benzoyl fluoride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B1115-25G
B1115-5G
TCB1115-25GEA
204
USD
TCB1115-25G
TCB1115-5G
Benzoyl fluoride ≥98.0% (by GC, titration analysis)
Benzoyl fluoride
CAS Number: 455-32-3
MDL Number: MFCD00000364
Molecular Formula: C7H5FO
Molecular Weight: 124.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 160
Melting point (°C): -28
Flash Point (°C): 49
Specific Gravity (20/20): 1.15
Formula:
C₇H₅FO Boiling Pt: 160 °C Melting Pt: -28 °C Density: 1.15 (20 °C) Flash Pt: 49 °C |
MDL Number:
MFCD00000364 CAS Number: 455-32-3 UN: 2920 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...