(S)-(-)-1,1'-Bi-2-naphthol ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
B1100-25G
B1100-5G
TCB1100-25GEA
305.5
USD
TCB1100-25G
TCB1100-5G
(S)-(-)-1,1'-Bi-2-naphthol ≥98.0% (by HPLC, titration analysis)
(S)-(-)-1,1'-Bi-2-naphthol
CAS Number: 18531-99-2
MDL Number: MFCD00004068
Molecular Formula: C20H14O2
Molecular Weight: 286.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 210
Specific rotation [a]20/D: -36 deg (C=1, THF)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Asymmetric Synthesis
Other Product Names / Synonyms: (S)-(-)-BINOL, (S)-(-)-2,2'-Dihydroxy-1,1'-binaphthyl
Formula:
C₂₀H₁₄O₂ MW: 286.33 g/mol Melting Pt: 210 °C |
MDL Number:
MFCD00004068 CAS Number: 18531-99-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...