4-tert-Butylcyclohexylamine ≥98.0% (by GC)

Supplier: TCI America

B1084-250ML B1084-25ML B1084-5ML
TCB1084-250MLEA 727 USD
TCB1084-250ML TCB1084-25ML TCB1084-5ML
4-tert-Butylcyclohexylamine ≥98.0% (by GC)
4-tert-Butylcyclohexylamine

CAS Number: 5400-88-4
MDL Number: MFCD00013763
Molecular Formula: C10H21N
Molecular Weight: 155.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 79
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: 4-Amino-1-tert-butylcyclohexane

Formula: C₁₀H₂₁N
Density: 0.87 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00013763
CAS Number: 5400-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a870c61c6d6460f70b51628114acbf1d1246eb25&vid=ab52a0ea818ab17473b04126fbec455ce57a9ad7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR