Toluene-α-sulfonyl fluoride ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride
B1062-25G
B1062-5G
TCB1062-25GEA
146.51
USD
TCB1062-25G
TCB1062-5G
Toluene-α-sulfonyl fluoride ≥98.0% (by GC)
Toluene-α-sulphonyl fluoride
CAS Number: 329-98-6
MDL Number: MFCD00007424
Molecular Formula: C7H7FO2S
Molecular Weight: 174.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 85
Melting point (°C): 94
Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: Phenylmethylsulfonyl Fluoride, PMSF
Formula:
C₇H₇FO₂S MW: 174.2 g/mol Boiling Pt: 85 °C Melting Pt: 94 °C |
MDL Number:
MFCD00007424 CAS Number: 329-98-6 UN: 2923 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...