(±)-3-Butyne-2-ol ≥97.0%

Supplier: TCI America

Synonyms: (±)-But-3-yn-2-ol, (±)-3-Hydroxy-1-butyne

B1001-25ML B1001-5ML
TCB1001-25MLEA 161.79 USD
TCB1001-25ML TCB1001-5ML
(±)-3-Butyne-2-ol ≥97.0%
(±)-3-Butyne-2-ol

CAS Number: 2028-63-9
MDL Number: MFCD00004541
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 34
Specific Gravity (20/20): 0.90


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 1-Butyn-3-ol, Ethynylmethylcarbinol

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 110 °C
Melting Pt: -12
Density: 0.90 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00004541
CAS Number: 2028-63-9
UN: 2929
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d3538a4cc333e7160ffa74384d22f84bd675a3b&vid=6d3538a4cc333e7160ffa74384d22f84bd675a3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR