tert-Butylchlorodimethylsilane ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride
[tert-Butyldimethylsilylating Agent]
CAS Number: 18162-48-6
MDL Number: MFCD00000501
Molecular Formula: C6H15ClSi
Molecular Weight: 150.72
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 88
Flash Point (°C): 28
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis, Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents, Bioscience, Reagents for Oligosaccharide Synthesis, Analytical Scien
Other Product Names / Synonyms: tert-Butylchlorodimethylsilane, tert-Butyldimethylsilyl Chloride, TBDMSCl, TBSCl
Formula:
C₆H₁₅ClSi MW: 150.72 g/mol Boiling Pt: 125 °C Melting Pt: 88 °C Flash Pt: 28 °C |
MDL Number:
MFCD00000501 CAS Number: 18162-48-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...