3-tert-Butyltoluene ≥98.0% (by GC)

Supplier: TCI America

B0967-1G B0967-5G
TCB0967-1GEA 146.5 USD
TCB0967-1G TCB0967-5G
3-tert-Butyltoluene ≥98.0% (by GC)
3-tert-Butyltoluene

CAS Number: 1075-38-3
MDL Number: MFCD00059210
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Melting point (°C): -41
Flash Point (°C): 38
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: 1-tert-Butyl-3-methylbenzene

Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 189 °C
Melting Pt: -41 °C
Density: 0.87 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00059210
CAS Number: 1075-38-3
UN: 2667
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02b8f47feb94676af6042da4736367261dead0ce&vid=02b8f47feb94676af6042da4736367261dead0ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR