(R)-(-)-2-Butanol ≥99.0% (by GC)

Supplier: TCI America

Synonyms: (R)-(-)-Ethyl methyl carbinol, (R)-(-)-sec-Butyl alcohol, (R)-(-)-2-Hydroxy butane, (R)-(-)-sec-Butanol, (R)-(-)-Methyl ethyl carbinol

B0926-1ML B0926-5ML
TCB0926-1MLEA 55 USD
TCB0926-1ML TCB0926-5ML
(R)-(-)-2-Butanol ≥99.0% (by GC)
(R)-(-)-2-Butanol

CAS Number: 14898-79-4
MDL Number: MFCD00064280
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 24
Specific Gravity (20/20): 0.81
Specific rotation [a]20/D: -13 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (R)-(-)-sec-Butyl Alcohol

Formula: C₄H₁₀O
MW: 74.12 g/mol
Boiling Pt: 100 °C
Density: 0.81 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00064280
CAS Number: 14898-79-4
UN: 1120
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75068adc0e890b6206e744ac10dac7fe5ae9842c&vid=75068adc0e890b6206e744ac10dac7fe5ae9842c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR